Dimethylcyanamide ≥98.0%

Supplier: TCI America

D1150-25ML
TCD1150-025MLEA 219.82 USD
TCD1150-025ML
Dimethylcyanamide ≥98.0%
Dimethylcyanamide

CAS Number: 1467-79-4
MDL Number: MFCD00001767
Molecular Formula: C3H6N2
Molecular Weight: 70.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 164
Flash Point (°C): 71
Specific Gravity (20/20): 0.89
Storage Temperature: 0-10°C

Formula: C₃H₆N₂
Boiling Pt: 164 °C
Density: 0.89 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00001767
CAS Number: 1467-79-4
https://us.vwr.com/stibo/web/std.lang.all/80/84/28978084.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR