Didecyl disulfide ≥95.0%
Supplier: TCI America
Synonyms:
1,2-Didecyldisulphane
, Decyl Disulphide
, Decyl Disulfide
, 1,2-Didecyldisulfane
D1141-10ML
TCD1141-010MLEA
191
USD
TCD1141-010ML
Didecyl disulfide ≥95.0%
Didecyl disulfide
CAS Number: 10496-18-1
MDL Number: MFCD00039859
Molecular Formula: C20H42S2
Molecular Weight: 346.68
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 208
Freezing point (°C): 17
Specific Gravity (20/20): 0.89
Other Product Names / Synonyms: Decyl Disulfide
Formula:
C₂₀H₄₂S₂ MW: 346.68 g/mol Boiling Pt: 208 °C Density: 0.89 (20 °C) |
MDL Number:
MFCD00039859 CAS Number: 10496-18-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...