2,2-Diisopropyl-1,3-dioxolane ≥96.0%
Supplier: TCI America
D1129-25G
TCD1129-025GEA
51
USD
TCD1129-025G
2,2-Diisopropyl-1,3-dioxolane ≥96.0%
2,2-Diisopropyl-1,3-dioxolane
CAS Number: 4421-10-7
MDL Number: MFCD00020902
Molecular Formula: C9H18O2
Molecular Weight: 158.24
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 172
Flash Point (°C): 48
Specific Gravity (20/20): 0.93
Formula:
C₉H₁₈O₂ Boiling Pt: 172 °C Density: 0.93 (20 °C) Flash Pt: 48 °C |
MDL Number:
MFCD00020902 CAS Number: 4421-10-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...