4,4'-Diamyloxyazoxybenzene ≥95.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
4,4'-Dipentyloxyazoxybenzene
D1093-1G
TCD1093-001GEA
406
USD
TCD1093-001G
4,4'-Diamyloxyazoxybenzene ≥95.0% (by total nitrogen basis)
4,4'-Diamyloxyazoxybenzene
CAS Number: 19482-05-4
MDL Number: MFCD00041933
Molecular Formula: C22H30N2O3
Molecular Weight: 370.49
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4,4'-Dipentyloxyazoxybenzene
Formula:
C₂₂H₃₀N₂O₃ MW: 370.49 g/mol |
MDL Number:
MFCD00041933 CAS Number: 19482-05-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...