3',4'-Dichlorosalicylanilide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

D1077-5G D1077-1G
TCD1077-005GEA 770.16 USD
TCD1077-005G TCD1077-1G
3',4'-Dichlorosalicylanilide ≥98.0% (by total nitrogen basis)
3',4'-Dichlorosalicylanilide

CAS Number: 24448-73-5
MDL Number: MFCD00020023
Molecular Formula: C13H9Cl2NO2
Molecular Weight: 282.12
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 214

Formula: C₁₃H₉Cl₂NO₂
Melting Pt: 214 °C
MDL Number: MFCD00020023
CAS Number: 24448-73-5
https://us.vwr.com/stibo/web/std.lang.all/63/11/28976311.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR