trans-1,3-Dimethylcyclohexane ≥95.0%

Supplier: TCI America

D1054-10ML
TCD1054-010MLEA 162.35 USD
TCD1054-010ML
trans-1,3-Dimethylcyclohexane ≥95.0%
trans-1,3-Dimethylcyclohexane

CAS Number: 2207-03-6
MDL Number: MFCD00064953
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Specific Gravity (20/20): 0.78


Other Product Names / Synonyms: trans-Hexahydro-m-xylene

Formula: C₈H₁₆
Boiling Pt: 124 °C
Density: 0.78 (20 °C)
MDL Number: MFCD00064953
CAS Number: 2207-03-6

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR