N-(2,4-Dinitrophenyl)-L-valine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-Dnp-L-valine
D1043-1G
D1043-100MG
TCD1043-001GEA
76.09
USD
TCD1043-001G
TCD1043-100MG
N-(2,4-Dinitrophenyl)-L-valine ≥98.0% (by HPLC, titration analysis)
N-(2,4-Dinitrophenyl)-L-valine
CAS Number: 1694-97-9
MDL Number: MFCD00038096
Molecular Formula: C11H13N3O6
Molecular Weight: 283.24
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 131
Specific rotation [a]20/D: -26 deg (C=1, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Dnp-L-valine, Dnp-Val-OH
Formula:
C₁₁H₁₃N₃O₆ MW: 283.24 g/mol Melting Pt: 131 °C |
MDL Number:
MFCD00038096 CAS Number: 1694-97-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...