N-(2,4-Dinitrophenyl)-DL-threonine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Dnp-DL-threonineDnp-DL-Thr-OH

D1039-100MG
TCD1039-100MGEA 47.69 USD
TCD1039-100MG
N-(2,4-Dinitrophenyl)-DL-threonine ≥99.0% (by titrimetric analysis)
N-(2,4-Dinitrophenyl)-DL-threonine

CAS Number: 14401-07-1
MDL Number: MFCD00070530
Molecular Formula: C10H11N3O7
Molecular Weight: 285.21
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 154


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Dnp-DL-threonine, Dnp-DL-Thr-OH

Formula: C₁₀H₁₁N₃O₇
MW: 285.21 g/mol
Melting Pt: 154 °C
MDL Number: MFCD00070530
CAS Number: 14401-07-1
https://us.vwr.com/stibo/web/std.lang.all/62/58/28976258.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR