N-(2,4-Dinitrophenyl)-L-serine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Dnp-L-serineDnp-Ser-OH

D1037-1G D1037-100MG
TCD1037-100MGEA 49.72 USD
TCD1037-100MG TCD1037-001G
N-(2,4-Dinitrophenyl)-L-serine ≥98.0% (by HPLC, titration analysis)
N-(2,4-Dinitrophenyl)-L-serine

CAS Number: 1655-64-7
MDL Number: MFCD00038098
Molecular Formula: C9H9N3O7
Molecular Weight: 271.19
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 177
Specific rotation [a]20/D: -23 deg (C=0.5, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Dnp-L-serine, Dnp-Ser-OH

Formula: C₉H₉N₃O₇
MW: 271.19 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00038098
CAS Number: 1655-64-7
https://us.vwr.com/stibo/web/std.lang.all/62/55/28976255.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR