Dodecylamine acetate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Aminododecane AcetateDodecylamineacetateLaurylamine Acetate

D0981-25G
TCD0981-025GEA 153.72 USD
TCD0981-025G
Dodecylamine acetate ≥98.0% (by titrimetric analysis)
Dodecylamine acetate

CAS Number: 2016-56-0
MDL Number: MFCD00043524
Molecular Formula: C12H27N
Molecular Weight: 245.41
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 69


Other Product Names / Synonyms: Aminododecane Acetate, Laurylamine Acetate

Formula: C₁₂H₂₇N
MW: 245.41 g/mol
Melting Pt: 69 °C
MDL Number: MFCD00043524
CAS Number: 2016-56-0
https://us.vwr.com/stibo/web/std.lang.all/61/59/28976159.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR