Diisopropylbenzene (mixture of isomers) ≥97.0%

Supplier: TCI America

D0929-25ML
TCD0929-025MLEA 34.49 USD
TCD0929-025ML
Diisopropylbenzene (mixture of isomers) ≥97.0%
Diisopropylbenzene (mixture of isomers)

CAS Number: 25321-09-9
MDL Number: MFCD00054263
Molecular Formula: C12H18
Molecular Weight: 162.28
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 77
Specific Gravity (20/20): 0.86

Formula: C₁₂H₁₈
Density: 0,86 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00054263
CAS Number: 25321-09-9

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR