Diphenylthiocarbazide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 1,5-Diphenyl-3-thiocarbohydrazide

D0917-5G
TCD0917-005GEA 119 USD
TCD0917-005G
Diphenylthiocarbazide ≥98.0% (by HPLC)
Diphenylthiocarbazide

CAS Number: 622-03-7
MDL Number: MFCD00046042
Molecular Formula: C13H14N4S
Molecular Weight: 258.34
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 155


Other Product Names / Synonyms: 1,5-Diphenyl-3-thiocarbohydrazide

Formula: C₁₃H₁₄N₄S
MW: 258.34 g/mol
Melting Pt: 155 °C
MDL Number: MFCD00046042
CAS Number: 622-03-7
https://us.vwr.com/stibo/web/std.lang.all/60/53/28976053.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR