Nitrosodiphenylamine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: DiphenylnitrosamineN-Nitrosodiphenylamine

D0899-25G D0899-500G
TCD0899-025GEA 29.51 USD
TCD0899-025G TCD0899-500G
Nitrosodiphenylamine ≥98.0% (by HPLC)
Nitrosodiphenylamine

CAS Number: 86-30-6
MDL Number: MFCD00019920
Molecular Formula: C12H10N2O
Molecular Weight: 198.23
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 68


Product Categories: Bioscience, ROS/RNS Generators, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: Diphenylnitrosamine

Formula: C₁₂H₁₀N₂O
MW: 198.22 g/mol
Melting Pt: 68 °C
MDL Number: MFCD00019920
CAS Number: 86-30-6
EINECS: 201-663-0
https://us.vwr.com/stibo/web/std.lang.all/60/23/28976023.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR