Benzil dioxime ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
Diphenylglyoxime
D0891-25G
TCD0891-025GEA
66.84
USD
TCD0891-025G
Benzil dioxime ≥98.0% (by total nitrogen basis)
Benzil dioxime
CAS Number: 23873-81-6
MDL Number: MFCD00002113
Molecular Formula: C14H12N2O2
Molecular Weight: 240.26
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 240
Other Product Names / Synonyms: Diphenylglyoxime
Formula:
C₁₄H₁₂N₂O₂ MW: 240.26 g/mol Melting Pt: 240 °C |
MDL Number:
MFCD00002113 CAS Number: 23873-81-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...