cis-6-Nonenyl acetate ≥92.0%

Supplier: TCI America

Synonyms: Acetic Acid cis-6-Nonenyl Ester

A0921-5ML
TCA0921-005MLEA 108.56 USD
TCA0921-005ML
cis-6-Nonenyl acetate ≥92.0%
cis-6-Nonenyl acetate

CAS Number: 76238-22-7
MDL Number: MFCD00059409
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: 92.0% (GC)
Form: Clear Liquid
Flash Point (°C): 106
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Acetic Acid cis-6-Nonenyl Ester

Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Density: 0.89 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD00059409
CAS Number: 76238-22-7

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR