1,5-Diphenylcarbazone 40,0% (by HPLC) (contains Diphenylcarbazide)

Supplier: TCI America

Synonyms: sym-DiphenylcarbazonePhenylazoformic acid 2-phenylhydrazidPhenylazoameisensäure-2-phenylhydrazidPhenylazoformic acid 2-phenylhydrazide

D0881-25G
TCD0881-025GEA 211.06 USD
TCD0881-025G
1,5-Diphenylcarbazone 40,0% (by HPLC) (contains Diphenylcarbazide)
1,5-Diphenylcarbazone

CAS Number: 538-62-5
MDL Number: MFCD00003024
Molecular Formula: C13H12N4O
Molecular Weight: 240.27
Purity/Analysis Method: 40.0% (HPLC)
Form: Crystal
Melting point (°C): 157

Formula: C₁₃H₁₂N₄O
MW: 240.26 g/mol
Boiling Pt: 440 °C (1013 hPa)
Melting Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00003024
CAS Number: 538-62-5
EINECS: 208-698-0
Merck Index: 12,03380
https://us.vwr.com/stibo/web/std.lang.all/59/97/28975997.jpg

Order Now

Specification Test Results

Purity/Analysis Method 40.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR