Girard's reagent D (N,N-dimethylglycine hydrazide dihydrochloride) ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Girard's Reagent DN,N-Dimethylglycine Hydrazide dihydrochloride

D0727-10G
TCD0727-010GEA 246.57 USD
TCD0727-010G
Girard's reagent D (N,N-dimethylglycine hydrazide dihydrochloride) ≥99.0% (by titrimetric analysis)
Girard's reagent D (N,N-Dimethylglycine hydrazide dihydrochloride)

CAS Number: 5787-71-3
MDL Number: MFCD00042020
Molecular Formula: C4H11N3O
Molecular Weight: 190.07
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 213


Other Product Names / Synonyms: N,N-Dimethylglycine Hydrazide Dihydrochloride

Formula: C₄H₁₃Cl₂N₃O
MW: 190.07 g/mol
Melting Pt: 213 °C
MDL Number: MFCD00042020
CAS Number: 5787-71-3
https://us.vwr.com/stibo/web/std.lang.all/57/04/28975704.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR