3,4-Dimethylcyclohexanol (mixture of isomers) ≥97.0%

Supplier: TCI America

Synonyms: 3,4-Dimethylcyclohexanol

D0703-25ML
TCD0703-025MLEA 179.58 USD
TCD0703-025ML
3,4-Dimethylcyclohexanol (mixture of isomers) ≥97.0%
3,4-Dimethylcyclohexanol (mixture of isomers)

CAS Number: 5715-23-1
MDL Number: MFCD00001506
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 78
Specific Gravity (20/20): 0.92

Formula: C₈H₁₆O
Boiling Pt: 189 °C
Density: 0.92 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00001506
CAS Number: 5715-23-1
https://us.vwr.com/stibo/web/std.lang.all/56/41/28975641.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR