1,3-Dimethylbutylamine ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-4-methylpentane

D0694-5ML
TCD0694-005MLEA 149.08 USD
TCD0694-005ML
1,3-Dimethylbutylamine ≥98.0%
1,3-Dimethylbutylamine

CAS Number: 108-09-8
MDL Number: MFCD00008088
Molecular Formula: C6H15N
Molecular Weight: 101.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 13
Specific Gravity (20/20): 0.75


Other Product Names / Synonyms: 2-Amino-4-methylpentane

Formula: C₆H₁₅N
MW: 101.19 g/mol
Boiling Pt: 108 °C
Density: 0.75 (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00008088
CAS Number: 108-09-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR