3-(Dimethylamino)propiononitrile ≥98.0%

Warning

Synonyms: 3-(Dimethylamino)propionitrile3-Dimethylaminopropionitrile3-(Dimethylamino)-Propionitrileβ-(Dimethylamino)propiononitrile

D0662-25ML
TCD0662-025MLEA 30.9 USD
TCD0662-025ML
3-(Dimethylamino)propiononitrile ≥98.0%
3-(Dimethylamino)propiononitrile

CAS Number: 1738-25-6
MDL Number: MFCD00001957
Molecular Formula: C5H10N2
Molecular Weight: 98.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Flash Point (°C): 64
Specific Gravity (20/20): 0.87

Formula: C₅H₁₀N₂
MW: 98.15 g/mol
Boiling Pt: 171 °C
Melting Pt: –45 °C
Density: 0.87 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00001957
CAS Number: 1738-25-6
EINECS: 217-090-4
https://us.vwr.com/stibo/web/std.lang.all/55/49/28975549.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR