Diisobutyl disulfide ≥98.0%
Supplier: TCI America
Synonyms:
Isobutyl Disulfide
, Isobutyl Disulphide
D0616-25ML
TCD0616-025MLEA
260
USD
TCD0616-025ML
Diisobutyl disulfide ≥98.0%
Diisobutyl disulfide
CAS Number: 1518-72-5
MDL Number: MFCD00039850
Molecular Formula: C8H18S2
Molecular Weight: 178.35
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 109
Specific Gravity (20/20): 0.93
Other Product Names / Synonyms: Isobutyl Disulfide
Formula:
C₈H₁₈S₂ MW: 178.35 g/mol Boiling Pt: 109 °C Density: 0.93 (20 °C) |
MDL Number:
MFCD00039850 CAS Number: 1518-72-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...