Diisobutylamine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D0615-25G
TCD0615-025GEA 87 USD
TCD0615-025G
Diisobutylamine hydrochloride ≥98.0% (by titrimetric analysis)
Diisobutylamine hydrochloride

CAS Number: 18251-82-6
MDL Number: MFCD00060151
Molecular Formula: C8H19N
Molecular Weight: 165.71
Purity/Analysis Method: 98.0% (T)
Form: Crystal

Formula: C₈H₁₉N
MDL Number: MFCD00060151
CAS Number: 18251-82-6
https://us.vwr.com/stibo/web/std.lang.all/54/19/28975419.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR