1,3-Dihydroxyacetone dimer ≥96.0% (by titrimetric analysis)
Synonyms:
2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol
D0560-25G
D0560-500G
D0560-100G
TCD0560-025GEA
24.72
USD
TCD0560-025G
TCD0560-100G
TCD0560-500G
1,3-Dihydroxyacetone dimer ≥96.0% (by titrimetric analysis)
1,3-Dihydroxyacetone dimer
CAS Number: 26776-70-5
MDL Number: MFCD00051019
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 80
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 80 °C |
MDL Number:
MFCD00051019 CAS Number: 26776-70-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...