Digitoxigenin ≥97.0% (by HPLC)

Supplier: TCI America

D0541-10MG
TCD0541-010MGEA 221.83 USD
TCD0541-010MG
Digitoxigenin ≥97.0% (by HPLC)
Digitoxigenin

CAS Number: 143-62-4
MDL Number: MFCD00003687
Molecular Formula: C23H34O4
Molecular Weight: 374.52
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal
Melting point (°C): 253
Specific rotation [a]20/D: 18 deg (C=1, MeOH)

Formula: C₂₃H₃₄O₄
MDL Number: MFCD00003687
CAS Number: 143-62-4

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR