Digitonin

Supplier: TCI America

D0540-100MG
TCD0540-100MGEA 103.51 USD
TCD0540-100MG
Digitonin
Digitonin

CAS Number: 11024-24-1
MDL Number: MFCD00077729
Molecular Formula: C56H92O29
Molecular Weight: 1229.32
Form: Crystal
Color: White
Melting point (°C): 240
Specific rotation [a]20/D: -52 deg (C=1, EtOH)

Formula: C₅₆H₉₂O₂₉
Melting Pt: 240 °C
MDL Number: MFCD00077729
CAS Number: 11024-24-1
UN: 3462
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/51/75/28975175.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR