N(α)-Acetyl-L-tryptophan ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
Ac-Trp-OH
, N-Acetyl-L-tryptophan
A0121-25G
A0121-1G
TCA0121-25GEA
82.62
USD
TCA0121-25G
TCA0121-001G
N(α)-Acetyl-L-tryptophan ≥98.0% (by HPLC)
N(α)-Acetyl-L-tryptophan
CAS Number: 1218-34-4
MDL Number: MFCD00065976
Molecular Formula: C13H14N2O3
Molecular Weight: 246.27
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 186
Specific rotation [a]20/D: 26 deg (C=1, 1mol/L NaOH sol.)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-Trp-OH
Formula:
C₁₃H₁₄N₂O₃ MW: 246.3 g/mol Melting Pt: 186 °C |
MDL Number:
MFCD00065976 CAS Number: 1218-34-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...