N,N-Diethylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Diethylcarbamyl chloride
D0482-25G
D0482-500G
D0482-100G
TCD0482-025GEA
24.5
USD
TCD0482-025G
TCD0482-500G
TCD0482-100G
N,N-Diethylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
N,N-Diethylcarbamoyl chloride
CAS Number: 88-10-8
MDL Number: MFCD00000636
Molecular Formula: C5H10ClNO
Molecular Weight: 135.59
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 76
Specific Gravity (20/20): 1.07
Formula:
C₅H₁₀ClNO Boiling Pt: 195 °C Density: 1.07 (20 °C) Flash Pt: 76 °C |
MDL Number:
MFCD00000636 CAS Number: 88-10-8 EINECS: 201-798-5 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...