α,α,α,2,4-Pentachlorotoluene ≥99.0%

Supplier: TCI America

Synonyms: 2,4-Dichlorobenzotrichloride

D0428-500G D0428-25G
TCD0428-500GEA 238.65 USD
TCD0428-500G TCD0428-025G
α,α,α,2,4-Pentachlorotoluene ≥99.0%
α,α,α,2,4-Pentachlorotoluene

CAS Number: 13014-18-1
MDL Number: MFCD00018818
Molecular Formula: C7H3Cl5
Molecular Weight: 264.35
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 159
Melting point (°C): 49


Other Product Names / Synonyms: alpha,alpha,alpha,2,4-Pentachlorotoluene

Formula: C₇H₃Cl₅
Boiling Pt: 159 °C
Melting Pt: 49 °C
MDL Number: MFCD00018818
CAS Number: 13014-18-1
https://us.vwr.com/stibo/web/std.lang.all/49/62/28974962.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR