2,4-Dichloro-1-naphthol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,3-Dichloro-4-hydroxynaphthalene

D0383-25G D0383-5G
TCD0383-25GEA 124.63 USD
TCD0383-25G TCD0383-005G
2,4-Dichloro-1-naphthol ≥97.0% (by GC, titration analysis)
2,4-Dichloro-1-naphthol

[for photography]
CAS Number: 2050-76-2
MDL Number: MFCD00003931
Molecular Formula: C10H6Cl2O
Molecular Weight: 213.06
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Melting point (°C): 105

Formula: C₁₀H₆Cl₂O
MW: 213.06 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 105 °C
MDL Number: MFCD00003931
CAS Number: 2050-76-2
EINECS: 218-103-6
https://us.vwr.com/stibo/web/std.lang.all/48/81/28974881.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR