TCIAD0379

D0379-1G
TCD0379-001GEA 193.7 USD
TCD0379-001G
TCIAD0379
[Redox Indicator]
CAS Number: 5415-26-9
MDL Number: MFCD00019446
Molecular Formula: C13H9Cl2NO2
Molecular Weight: 304.10
Form: Crystal
Color: Purple

Other Product Names / Synonyms: 2,6-Dichlorophenol-indo-o-cresol Sodium Salt
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR