TOKYD0367

D0367-25G
TCD0367-025GEA 182.53 USD
TCD0367-025G
TOKYD0367
(stabilized with MEHQ)
CAS Number: 156-59-2
MDL Number: MFCD00000925
Molecular Formula: C2H2Cl2
Molecular Weight: 96.94
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 61
Flash Point (°C): 4
Specific Gravity (20/20): 1.29

Other Product Names / Synonyms: cis-Acetylene Dichloride
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR