1,2-Dichloro-1-ethoxyethane ≥93.0%

Supplier: TCI America

Synonyms: 1,2-Dichloroethyl ethyl ether1,2-Dichlorodiethyl ether

D0355-25G
TCD0355-025GEA 349.01 USD
TCD0355-025G
1,2-Dichloro-1-ethoxyethane ≥93.0%
1,2-Dichloro-1-ethoxyethane

CAS Number: 623-46-1
MDL Number: MFCD00041730
Molecular Formula: C4H8Cl2O
Molecular Weight: 143.01
Purity/Analysis Method: 93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 35
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: 1,2-Dichlorodiethyl Ether

Formula: H₃CCH₂OCClHCClH₂
MW: 143.01 g/mol
Boiling Pt: 145 °C
Melting Pt: –72 °C
Density: 1.17 (20 °C)
Flash Pt: 35 °C
MDL Number: MFCD00041730
CAS Number: 623-46-1
EINECS: 210-794-2
UN: 1993
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/48/48/28974848.jpg

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR