TCIAD0354

D0354-25ML
TCD0354-025MLEA 170.71 USD
TCD0354-025ML
TCIAD0354
CAS Number: 619-33-0
MDL Number: MFCD00000849
Molecular Formula: C6H12Cl2O2
Molecular Weight: 187.06
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 184
Flash Point (°C): 59
Specific Gravity (20/20): 1.15

Other Product Names / Synonyms: Dichloroacetaldehyde Diethyl Acetal, 2,2-Dichloro-1,1-diethoxyethane
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR