Dichloroacetaldehyde hydrate ≥90.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,2-Dichloro-1,1-ethanediol
D0312-25G
TCD0312-025GEA
626.82
USD
TCD0312-025G
Dichloroacetaldehyde hydrate ≥90.0% (by titrimetric analysis)
Dichloroacetaldehyde hydrate
CAS Number: 16086-14-9
MDL Number: MFCD00150652
Molecular Formula: C2H2Cl2O
Molecular Weight: 112.94
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 121
Melting point (°C): 56
Other Product Names / Synonyms: 2,2-Dichloro-1,1-ethanediol
Formula:
C₂H₂Cl₂O MW: 112.94 g/mol Boiling Pt: 121 °C Melting Pt: 56 °C |
MDL Number:
MFCD00150652 CAS Number: 16086-14-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...