Dibutyl phosphonate ≥95.0%
Supplier: TCI America
Synonyms:
Dibutyl phosphite
, Di-n-butylphosphite
CAS Number: 1809-19-4
MDL Number: MFCD00066633
Molecular Formula: C8H19O3P
Molecular Weight: 194.21
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 123
Specific Gravity (20/20): 0.99
Product Categories: Synthetic Organic Chemistry, Dialkyl Phosphites (Multi-component Reaction)
Other Product Names / Synonyms: Phosphonic Acid Dibutyl Ester, Dibutyl Phosphonate, Phosphorous Acid Dibutyl Ester
Formula:
C₈H₁₉O₃P MW: 194.21 g/mol Boiling Pt: 121 °C Density: 0.99 (20 °C) Flash Pt: 123 °C |
MDL Number:
MFCD00066633 CAS Number: 1809-19-4 EINECS: 217-316-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...