4-Chloromethyl-o-xylene ≥70.0% (contains isomer)
Supplier: TCI America
Synonyms:
α-Chloro-1,3,4-trimethylbenzene
, 1-(Chloromethyl)-3,4-dimethylbenzene
, 4-(Chloromethyl)-1,2-dimethylbenzene
, 3,4-Dimethylbenzyl chloride
D0290-25G
TCD0290-025GEA
166.7
USD
TCD0290-025G
4-Chloromethyl-o-xylene ≥70.0% (contains isomer)
4-Chloromethyl-o-xylene
CAS Number: 102-46-5
MDL Number: MFCD00000910
Molecular Formula: C9H11Cl
Molecular Weight: 154.64
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 96
Specific Gravity (20/20): 1.06
Other Product Names / Synonyms: 1-(Chloromethyl)-3,4-dimethylbenzene
Formula:
C₉H₁₁Cl MW: 154.64 g/mol Boiling Pt: 117 °C Density: 1.06 (20 °C) Flash Pt: 96 °C |
MDL Number:
MFCD00000910 CAS Number: 102-46-5 EINECS: 203-032-5 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...