Direct blue 6
Supplier: TCI America
Synonyms:
Nippon Blue B
, Benzo Blue 2B
D0235-1G
TCD0235-001GEA
82.18
USD
TCD0235-001G
Direct blue 6
Direct blue 6
CAS Number: 2602-46-2
MDL Number: MFCD00068426
Molecular Formula: C32H24N6O14S4
Molecular Weight: 932.74
Form: Crystal
Color: Gray
Other Product Names / Synonyms: Benzo Blue 2B, Nippon Blue B
Formula:
C₃₂H₂₄N₆O₁₄S₄ MW: 932.74 g/mol |
MDL Number:
MFCD00068426 CAS Number: 2602-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...