Direct scarlet B

Supplier: TCI America

Synonyms: Diamine Scarlet BAmidine Scarlet BChloramine Red BPontamine Scarlet BDirect Red 37

D0234-25G
TCD0234-025GEA 102.48 USD
TCD0234-025G
Direct scarlet B
Direct scarlet B

CAS Number: 3530-19-6
MDL Number: MFCD00059531
Molecular Formula: C30H24N4O8S2
Molecular Weight: 676.63
Form: Crystal
Color: Yellow Red


Other Product Names / Synonyms: Amidine Scarlet B, Chloramine Red B, Diamine Scarlet B, Pontamine Scarlet B, Direct Red 37

Formula: C₃₀H₂₄N₄O₈S₂
MW: 676.63 g/mol
MDL Number: MFCD00059531
CAS Number: 3530-19-6
https://us.vwr.com/stibo/web/std.lang.all/46/65/28974665.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR