3,5-Dibromosulfanilamide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Amino-3,5-dibromobenzenesulphonamide4-Amino-3,5-dibromobenzenesulfonamide

D0212-25G
TCD0212-025GEA 311 USD
TCD0212-025G
3,5-Dibromosulfanilamide ≥97.0% (by titrimetric analysis)
3,5-Dibromosulphanilamide

CAS Number: 39150-45-3
MDL Number: MFCD00014783
Molecular Formula: C6H6Br2N2O2S
Molecular Weight: 329.99
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 242

https://us.vwr.com/stibo/web/std.lang.all/46/31/28974631.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR