N-Allylaniline ≥99.0%
Supplier: TCI America
A0880-500ML
A0880-25ML
A0880-100ML
TCA0880-025MLEA
56.84
USD
TCA0880-025ML
TCA0880-500ML
TCA0880-100ML
N-Allylaniline ≥99.0%
N-Allylaniline
CAS Number: 589-09-3
MDL Number: MFCD00008638
Molecular Formula: C9H11N
Molecular Weight: 133.19
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 89
Specific Gravity (20/20): 0.97
Formula:
C₉H₁₁N MW: 133.19 g/mol Boiling Pt: 220 °C Density: 0.97 (20 °C) Flash Pt: 89 °C |
MDL Number:
MFCD00008638 CAS Number: 589-09-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...