1,1-Dibromoethane ≥98.0% stabilized

Supplier: TCI America

Synonyms: Ethylidene bromide

D0184-5G
TCD0184-005GEA 130.89 USD
TCD0184-005G
1,1-Dibromoethane ≥98.0% stabilized
1,1-Dibromoethane

(stabilized with Copper chip)
CAS Number: 557-91-5
MDL Number: MFCD00041719
Molecular Formula: C2H4Br2
Molecular Weight: 187.86
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Specific Gravity (20/20): 2.10


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Ethylidene Bromide

Formula: Br₂HCCH₃
MW: 187.86 g/mol
Boiling Pt: 108 °C
Melting Pt: –0.5 °C
Density: 2.10 (20 °C)
MDL Number: MFCD00041719
CAS Number: 557-91-5
EINECS: 209-184-9
UN: 2810
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/45/87/28974587.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR