Dibenzoyl thiamine ≥98.0% (by titrimetric analysis)

Synonyms: 4-(N-((4-Amino-2-methylpyrimidin-5-yl)methyl)formamido)-3-(benzoylthio)pent-3-en-1-yl benzoate

D0151-1G D0151-25G
TCD0151-025GEA 176.49 USD
TCD0151-025G TCD0151-001G
Dibenzoyl thiamine ≥98.0% (by titrimetric analysis)
Dibenzoyl thiamine

CAS Number: 299-88-7
MDL Number: MFCD00867787
Molecular Formula: C26H26N4O4S
Molecular Weight: 490.58
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 173

Formula: C₂₆H₂₆N₄O₄S
MW: 490.58 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00867787
CAS Number: 299-88-7
https://us.vwr.com/stibo/web/std.lang.all/45/40/28974540.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR