N,N-Diacetyl-o-aminoazotoluene ≥90.0% (by HPLC)

Supplier: TCI America

Synonyms: Pellidol

D0063-25G
TCD0063-025GEA 215 USD
TCD0063-025G
N,N-Diacetyl-o-aminoazotoluene ≥90.0% (by HPLC)
N,N-Diacetyl-o-aminoazotoluene

CAS Number: 83-63-6
MDL Number: MFCD00026179
Molecular Formula: C18H19N3O2
Molecular Weight: 309.37
Purity/Analysis Method: 90.0% (HPLC)
Form: Crystal
Melting point (°C): 76


Other Product Names / Synonyms: Pellidol

Formula: C₁₈H₁₉N₃O₂
MW: 309.37 g/mol
Melting Pt: 76 °C
MDL Number: MFCD00026179
CAS Number: 83-63-6
https://us.vwr.com/stibo/web/std.lang.all/44/01/28974401.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR