Decenylsuccinic Anhydride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

D0030-25G
TCD0030-025GEA 82.18 USD
TCD0030-025G
Decenylsuccinic Anhydride ≥95.0% (by titrimetric analysis)
Decenylsuccinic anhydride

CAS Number: 25447-83-0
MDL Number: MFCD00014547
Molecular Formula: C14H22O3
Molecular Weight: 238.33
Purity/Analysis Method: 95.0% (T)
Form: Clear Liquid
Flash Point (°C): 174
Specific Gravity (20/20): 1.00

Formula: C₁₄H₂₂O₃
Density: 1,00 (20 °C)
Flash Pt: 174 °C
MDL Number: MFCD00014547
CAS Number: 25447-83-0

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR