Pentyl decanoate ≥93.0%
Supplier: TCI America
D0018-25ML
TCD0018-025MLEA
62
USD
TCD0018-025ML
Pentyl decanoate ≥93.0%
Pentyl decanoate
CAS Number: 5933-87-9
MDL Number: MFCD00059454
Molecular Formula: C15H30O2
Molecular Weight: 242.40
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: Decanoic Acid Amyl Ester, Pentyl Decanoate, Decanoic Acid Pentyl Ester
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...