(+/-)-cis-3-(Carbobenzoxyamino)cyclohexanecarboxylic acid ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (+/-)-cis-3-(Cbz-amino)cyclohexanecarboxylic Acid(1R,3S)-rel-3-(((Benzyloxy)carbonyl)amino)cyclohexanecarboxylic acid

C2397-1G
TCC2397-1GEA 261 USD
TCC2397-1G
(+/-)-cis-3-(Carbobenzoxyamino)cyclohexanecarboxylic acid ≥95.0% (by HPLC, titration analysis)
(+/-)-cis-3-(Carbobenzoxyamino)cyclohexanecarboxylic acid

CAS Number: 952616-39-6
Molecular Formula: C15H19NO4
Molecular Weight: 277.32
Purity/Analysis Method: 95.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: (+/-)-cis-3-(Cbz-amino)cyclohexanecarboxylic Acid

Formula: C₁₅H₁₉NO₄
MW: 277.32 g/mol
CAS Number: 952616-39-6
https://us.vwr.com/stibo/web/std.lang.all/93/76/28939376.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR