9-Chlorohexadecafluorononanoic acid ≥98.0% (by titrimetric analysis)

Synonyms: 9-Chloroperfluorononanoic acid

C2355-5G
TCC2355-5GEA 221.34 USD
TCC2355-5G
9-Chlorohexadecafluorononanoic acid ≥98.0% (by titrimetric analysis)
9-Chlorohexadecafluorononanoic acid

CAS Number: 865-79-2
MDL Number: MFCD00155671
Molecular Formula: C9HClF16O2
Molecular Weight: 480.53
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 79

Formula: C₉HClF₁₆O₂
MW: 480.53 g/mol
Boiling Pt: 130 °C
Melting Pt: 79 °C
MDL Number: MFCD00155671
CAS Number: 865-79-2
https://us.vwr.com/stibo/web/std.lang.all/93/48/28939348.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR