1,3,5-Cyclohexanetricarbonitrile (cis and trans mixture) ≥98.0%

Supplier: TCI America

Synonyms: 1,3,5-Tricyanocyclohexane

C2327-1G
TCC2327-1GEA 255.67 USD
TCC2327-1G
1,3,5-Cyclohexanetricarbonitrile (cis and trans mixture) ≥98.0%
1,3,5-Cyclohexanetricarbonitrile (cis and trans mixture)

CAS Number: 183582-92-5
MDL Number: MFCD08276327
Molecular Formula: C9H9N3
Molecular Weight: 159.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 200


Other Product Names / Synonyms: 1,3,5-Tricyanocyclohexane

Formula: C₉H₉N₃
MW: 159.19 g/mol
Boiling Pt: 200 °C
MDL Number: MFCD08276327
CAS Number: 183582-92-5
https://us.vwr.com/stibo/web/std.lang.all/93/28/28939328.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR