Coumarin 314 ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
Ethyl 2,3,6,7-Tetrahydro-11-oxo-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizine-10-carboxylate
, Ethyl 11-oxo-2,3,5,6,7,11-hexahydro-1H-pyrano[2,3-f]pyrido[3,2,1-ij]quinoline-10-carboxylate
, Coumarin 504
CAS Number: 55804-66-5
MDL Number: MFCD00051334
Molecular Formula: C18H19NO4
Molecular Weight: 313.35
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 150
Lambda max.: 436 nm (EtOH)
Product Categories: Materials Chemistry, Coumarin Dyes (Functional Organic Dyes), Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: Coumarin 504, Ethyl 2,3,6,7-Tetrahydro-11-oxo-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizine-10-carboxylate
Formula:
C₁₈H₁₉NO₄ MW: 313.35 g/mol Melting Pt: 150 °C |
MDL Number:
MFCD00051334 CAS Number: 55804-66-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...