Chlorothiazide ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
6-Chloro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-Dioxide
, 6-Chloro-2H-1,2,4-benzothiadiazine-7-sulphonamide 1,1-Dioxide
CAS Number: 58-94-6
MDL Number: MFCD00058576
Molecular Formula: C7H6ClN3O4S2
Molecular Weight: 295.71
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 348
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 6-Chloro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-Dioxide
Formula:
C₇H₆ClN₃O₄S₂ MW: 295.71 g/mol Melting Pt: 348 °C |
MDL Number:
MFCD00058576 CAS Number: 58-94-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...